Tisk…

Anti-TGM4 Mouse Monoclonal Antibody

Dodavatel: Biorbyt
BIRBORB22794-5EA 0 CZK
BIRBORB22794-5 BIRBORB22794-100 BIRBORB22794-20
Anti-TGM4 Mouse Monoclonal Antibody
Protilátky
Type: Primary
Antigen: TGM4 (transglutaminase 4)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b (Mouse)
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR