Tisk…

Anti-UBE2L6 Mouse Monoclonal Antibody

Dodavatel: Biorbyt
BIRBORB22795-5EA 0 CZK
BIRBORB22795-5 BIRBORB22795-100 BIRBORB22795-20
Anti-UBE2L6 Mouse Monoclonal Antibody
Protilátky
Type: Primary
Antigen: UBE2L6 (ubiquitin-conjugating enzyme E2L 6)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b (Mouse)
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR