Tisk…

Anti-YWHAB Mouse Monoclonal Antibody

Dodavatel: Biorbyt
BIRBORB22799-5EA 0 CZK
BIRBORB22799-5 BIRBORB22799-20 BIRBORB22799-100
Anti-YWHAB Mouse Monoclonal Antibody
Protilátky
Type: Primary
Antigen: YWHAB (tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, beta)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a (Mouse)
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR