Tisk…

Anti-SNCA Mouse Monoclonal Antibody

Dodavatel: Biorbyt
BIRBORB22805-100EA 0 CZK
BIRBORB22805-100 BIRBORB22805-20 BIRBORB22805-5
Anti-SNCA Mouse Monoclonal Antibody
Protilátky
Type: Primary
Antigen: SNCA (synuclein, alpha (non A4 component of amyloid precursor))
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 (Mouse)
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR