Tisk…

Anti-BID Mouse monoclonal antibody

Dodavatel: Biorbyt
BIRBORB22806-20EA 0 CZK
BIRBORB22806-20 BIRBORB22806-100 BIRBORB22806-5
Anti-BID Mouse monoclonal antibody
Protilátky
Type: Primary
Antigen: BID (BH3 interacting domain death agonist)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 (Mouse)
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR