Tisk…

Anti-CRABP2 Mouse Monoclonal Antibody

Dodavatel: Biorbyt
BIRBORB22816-20EA 0 CZK
BIRBORB22816-20 BIRBORB22816-100 BIRBORB22816-5
Anti-CRABP2 Mouse Monoclonal Antibody
Protilátky
Type: Primary
Antigen: CRABP2 (cellular retinoic acid binding protein 2)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a (Mouse)
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR