Tisk…
Anti-FABP1 Mouse monoclonal antibody
Dodavatel: Biorbyt
BIRBORB22822-20EA 0 CZK
BIRBORB22822-20 BIRBORB22822-5 BIRBORB22822-100
Anti-FABP1 Mouse monoclonal antibody
Protilátky
Type: Primary
Antigen: FABP1 (fatty acid binding protein 1, liver)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 (Mouse)
Reactivity: Human