Tisk…
Anti-FADD Mouse Monoclonal Antibody
Dodavatel: Biorbyt
BIRBORB22825-20EA 0 CZK
BIRBORB22825-20 BIRBORB22825-5 BIRBORB22825-100
Anti-FADD Mouse Monoclonal Antibody
Protilátky
Type: Primary
Antigen: FADD (Fas (TNFRSF6)-associated via death domain)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b (Mouse)
Reactivity: Human