Tisk…

Anti-GPS2 Mouse Monoclonal Antibody

Dodavatel: Biorbyt
BIRBORB22829-100EA 0 CZK
BIRBORB22829-100 BIRBORB22829-20 BIRBORB22829-5
Anti-GPS2 Mouse Monoclonal Antibody
Protilátky
Type: Primary
Antigen: GPS2 (G protein pathway suppressor 2)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG3 Kappa (Mouse)
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR