Tisk…

Anti-SNCG Rabbit Polyclonal Antibody

Dodavatel: Biorbyt
BIRBORB22830-5EA 0 CZK
BIRBORB22830-5 BIRBORB22830-100 BIRBORB22830-20
Anti-SNCG Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: SNCG (synuclein, gamma (breast cancer-specific protein 1))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR