Tisk…

Anti-MYC Mouse Monoclonal Antibody

Dodavatel: Biorbyt
BIRBORB22836-0.2EA 0 CZK
BIRBORB22836-0.2 BIRBORB22836-0.5 BIRBORB22836-100
Anti-MYC Mouse Monoclonal Antibody
Protilátky
Type: Primary
Antigen: MYC (v-myc avian myelocytomatosis viral oncogene homolog)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 (Mouse)
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR