Tisk…

Anti-MYC Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase))

Dodavatel: Biorbyt
BIRBORB22837-100EA 0 CZK
BIRBORB22837-100 BIRBORB22837-200
Anti-MYC Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase))
Protilátky
Type: Primary
Antigen: MYC (v-myc avian myelocytomatosis viral oncogene homolog)
Clonality: monoclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Mouse
Isotype: IgG1 (Mouse)
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR