Tisk…

Anti-KIR2DL3 Mouse Monoclonal Antibody

Dodavatel: Biorbyt
BIRBORB22852-5EA 0 CZK
BIRBORB22852-5 BIRBORB22852-20 BIRBORB22852-100
Anti-KIR2DL3 Mouse Monoclonal Antibody
Protilátky
Type: Primary
Antigen: KIR2DL3 (killer cell immunoglobulin-like receptor, two domains, long cytoplasmic tail, 3)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a (Mouse)
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR