Tisk…

Anti-MHC Class II (I-E) Mouse Monoclonal Antibody (Biotin)

Dodavatel: Biorbyt
BIRBORB22862-1EA 0 CZK
BIRBORB22862-1
Anti-MHC Class II (I-E) Mouse Monoclonal Antibody (Biotin)
Protilátky
Type: Primary
Antigen: MHC Class II (I-E)
Clonality: monoclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG2a (Mouse)
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR