Tisk…

Anti-HBsAg None antibody

Dodavatel: Biorbyt
BIRBORB22870-10EA 0 CZK
BIRBORB22870-10 BIRBORB22870-2
Anti-HBsAg None antibody
Protilátky
Type: Primary
Antigen: HBsAg
Clonality:
Clone:
Conjugation: unconjugated
Epitope:
Host: None
Isotype: IgG
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR