Tisk…

Anti-FOLH1 Mouse monoclonal antibody

Dodavatel: Biorbyt
BIRBORB22903-100EA 0 CZK
BIRBORB22903-100
Anti-FOLH1 Mouse monoclonal antibody
Protilátky
Type: Primary
Antigen: FOLH1 (folate hydrolase (prostate-specific membrane antigen) 1)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 (Mouse)
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR