Tisk…

Anti-REXO1 Mouse Monoclonal Antibody

Dodavatel: Biorbyt
BIRBORB22906-5EA 0 CZK
BIRBORB22906-5 BIRBORB22906-20 BIRBORB22906-100
Anti-REXO1 Mouse Monoclonal Antibody
Protilátky
Type: Primary
Antigen: REXO1 (REX1, RNA exonuclease 1 homolog (S. cerevisiae))
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b (Mouse)
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR