Tisk…

Anti-TARDBP Mouse Monoclonal Antibody

Dodavatel: Biorbyt
BIRBORB22910-100EA 0 CZK
BIRBORB22910-100 BIRBORB22910-5 BIRBORB22910-20
Anti-TARDBP Mouse Monoclonal Antibody
Protilátky
Type: Primary
Antigen: TARDBP (TAR DNA binding protein)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 (Mouse)
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR