Tisk…

Anti-HBsAg (ad/ay) Horse Polyclonal Antibody

Dodavatel: Biorbyt
BIRBORB23045-1EA 0 CZK
BIRBORB23045-1
Anti-HBsAg (ad/ay) Horse Polyclonal Antibody
Protilátky
Type: Primary
Antigen: HBsAg (ad/ay)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Horse
Isotype: none
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR