Tisk…

Anti-Strep pneumoniae None Polyclonal Antibody

Dodavatel: Biorbyt
BIRBORB23064-1EA 0 CZK
BIRBORB23064-1
Anti-Strep pneumoniae None Polyclonal Antibody
Protilátky
Type: Primary
Antigen: Strep pneumoniae
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: None
Isotype: none
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR