Tisk…

Anti-Herpes simplex Type-1 Goat Polyclonal Antibody

Dodavatel: Biorbyt
BIRBORB23101-1EA 0 CZK
BIRBORB23101-1
Anti-Herpes simplex Type-1 Goat Polyclonal Antibody
Protilátky
Type: Primary
Antigen: Herpes simplex Type-1
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR