Tisk…
Anti-Herpes simplex Type-1 Goat Polyclonal Antibody (FITC (Fluorescein))
Dodavatel: Biorbyt
BIRBORB23102-1EA 0 CZK
BIRBORB23102-1
Anti-Herpes simplex Type-1 Goat Polyclonal Antibody (FITC (Fluorescein))
Protilátky
Type: Primary
Antigen: Herpes simplex Type-1
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Goat
Isotype: none
Reactivity:
Objednat