Tisk…

Anti-HCV Core Goat Polyclonal Antibody

Dodavatel: Biorbyt
BIRBORB23116-1EA 0 CZK
BIRBORB23116-1
Anti-HCV Core Goat Polyclonal Antibody
Protilátky
Type: Primary
Antigen: HCV Core
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR