Tisk…

Anti-Treponema pallidum Rabbit Polyclonal Antibody (Biotin)

Dodavatel: Biorbyt
BIRBORB23128-1EA 0 CZK
BIRBORB23128-1
Anti-Treponema pallidum Rabbit Polyclonal Antibody (Biotin)
Protilátky
Type: Primary
Antigen: Treponema pallidum
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR