Tisk…
Anti-Rotavirus NCDV Goat Polyclonal Antibody (Biotin)
Dodavatel: Biorbyt
BIRBORB23132-1EA 0 CZK
BIRBORB23132-1
Anti-Rotavirus NCDV Goat Polyclonal Antibody (Biotin)
Protilátky
Type: Primary
Antigen: Rotavirus NCDV
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype: none
Reactivity:
Objednat