Tisk…

Anti-Rotavirus NCDV Goat Polyclonal Antibody (FITC (Fluorescein))

Dodavatel: Biorbyt
BIRBORB23133-1EA 0 CZK
BIRBORB23133-1
Anti-Rotavirus NCDV Goat Polyclonal Antibody (FITC (Fluorescein))
Protilátky
Immunofluorescence: 1:10-1:50

Type: Primary
Antigen: Rotavirus NCDV
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Goat
Isotype: none
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR