Tisk…

Anti-N. meningitidis Rabbit Polyclonal Antibody (FITC (Fluorescein))

Dodavatel: Biorbyt
BIRBORB23205-1EA 0 CZK
BIRBORB23205-1
Anti-N. meningitidis Rabbit Polyclonal Antibody (FITC (Fluorescein))
Protilátky
Type: Primary
Antigen: N. meningitidis
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR