Tisk…

Anti-Diphtheria toxin/toxoid Goat Polyclonal Antibody (Biotin)

Dodavatel: Biorbyt
BIRBORB23218-1EA 0 CZK
BIRBORB23218-1
Anti-Diphtheria toxin/toxoid Goat Polyclonal Antibody (Biotin)
Protilátky
Type: Primary
Antigen: Diphtheria toxin/toxoid
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype: none
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR