Tisk…

Anti-HBsAg (ad/ay) Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Dodavatel: Biorbyt
BIRBORB23237-1EA 0 CZK
BIRBORB23237-1
Anti-HBsAg (ad/ay) Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Protilátky
Type: Primary
Antigen: HBsAg (ad/ay)
Clonality: polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR