Tisk…
Anti-West Nile Virus Rabbit Polyclonal Antibody
Dodavatel: Biorbyt
BIRBORB23279-100EA 0 CZK
BIRBORB23279-100
Anti-West Nile Virus Rabbit Polyclonal Antibody
Protilátky
ELISA: 10 ng
Type: Primary
Antigen: West Nile Virus
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Objednat