Tisk…

Anti-SARS-CoV, Spike Rabbit Polyclonal Antibody

Dodavatel: Biorbyt
BIRBORB23297-100EA 0 CZK
BIRBORB23297-100
Anti-SARS-CoV, Spike Rabbit Polyclonal Antibody
Protilátky
ELISA: 10 ng

Type: Primary
Antigen: SARS-CoV, Spike
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR