Tisk…

Anti-B. anthracis LF Rabbit Polyclonal Antibody

Dodavatel: Biorbyt
BIRBORB23301-100EA 0 CZK
BIRBORB23301-100
Anti-B. anthracis LF Rabbit Polyclonal Antibody
Protilátky
ELISA: 10 ng

Type: Primary
Antigen: B. anthracis LF (Bacillus anthracis lethal factor)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR