Tisk…

Anti-C. botulinum Toxin B Mouse Monoclonal Antibody

Dodavatel: Biorbyt
BIRBORB23431-1EA 0 CZK
BIRBORB23431-1
Anti-C. botulinum Toxin B Mouse Monoclonal Antibody
Protilátky
  • Immunofluorescence: 1:25

Type: Primary
Antigen: C. botulinum Toxin B
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR