Tisk…

Anti-IBV Mouse Monoclonal Antibody

Dodavatel: Biorbyt
BIRBORB23540-1EA 0 CZK
BIRBORB23540-1
Anti-IBV Mouse Monoclonal Antibody
Protilátky
Type: Primary
Antigen: IBV
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR