Tisk…

Anti-HPV 6 (E7) Mouse Monoclonal Antibody

Dodavatel: Biorbyt
BIRBORB23582-1EA 0 CZK
BIRBORB23582-1
Anti-HPV 6 (E7) Mouse Monoclonal Antibody
Protilátky
Type: Primary
Antigen: HPV 6 (E7)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG3
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR