Tisk…

Anti-Chicken anti-Bovine (TSH) Chicken Polyclonal Antibody

Dodavatel: Biorbyt
BIRBORB23675-1EA 0 CZK
BIRBORB23675-1
Anti-Chicken anti-Bovine (TSH) Chicken Polyclonal Antibody
Protilátky
Type: Primary
Antigen: Chicken anti-Bovine (TSH)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Chicken
Isotype: none
Reactivity: Bovine
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR