Tisk…

Anti-XIAP Rabbit Polyclonal Antibody

Dodavatel: Biorbyt
BIRBORB23676-100EA 0 CZK
BIRBORB23676-100
Anti-XIAP Rabbit Polyclonal Antibody
Protilátky
  • Western Blot: 1:1000

Type: Primary
Antigen: XIAP (X-linked inhibitor of apoptosis)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR