Tisk…

Anti-Influenza B Neuraminidase Mouse Monoclonal Antibody

Dodavatel: Biorbyt
BIRBORB23729-0.5EA 0 CZK
BIRBORB23729-0.5
Anti-Influenza B Neuraminidase Mouse Monoclonal Antibody
Protilátky
Type: Primary
Antigen: Influenza B Neuraminidase
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR