Tisk…

Anti-BSA Chicken Polyclonal Antibody

Dodavatel: Biorbyt
BIRBORB23734-1EA 0 CZK
BIRBORB23734-1
Anti-BSA Chicken Polyclonal Antibody
Protilátky
Type: Primary
Antigen: BSA
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Chicken
Isotype: none
Reactivity: Bovine
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR