Tisk…

Anti-Influenza B Virus Mouse Monoclonal Antibody (FITC (Fluorescein))

Dodavatel: Biorbyt
BIRBORB23775-100EA 0 CZK
BIRBORB23775-100
Anti-Influenza B Virus Mouse Monoclonal Antibody (FITC (Fluorescein))
Protilátky
Type: Primary
Antigen: Influenza B Virus
Clonality: monoclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR