Tisk…

Anti-Verotoxin II (SLT-2b, STX-2b) Mouse Monoclonal Antibody

Dodavatel: Biorbyt
BIRBORB23857-100EA 0 CZK
BIRBORB23857-100
Anti-Verotoxin II (SLT-2b, STX-2b) Mouse Monoclonal Antibody
Protilátky
Type: Primary
Antigen: Verotoxin II (SLT-2b, STX-2b)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR