Tisk…
Anti-HPV 18 (E7) Mouse Monoclonal Antibody
Dodavatel: Biorbyt
BIRBORB23996-1EA 0 CZK
BIRBORB23996-1
Anti-HPV 18 (E7) Mouse Monoclonal Antibody
Protilátky
Type: Primary
Antigen: HPV 18 (E7)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Objednat