Tisk…
Anti-E. coli Enterotoxin B Mouse Monoclonal Antibody
Dodavatel: Biorbyt
BIRBORB24001-1EA 0 CZK
BIRBORB24001-1
Anti-E. coli Enterotoxin B Mouse Monoclonal Antibody
Protilátky
Type: Primary
Antigen: E. coli Enterotoxin B
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Objednat