Tisk…

Anti-Influenza A H5 Mouse Monoclonal Antibody

Dodavatel: Biorbyt
BIRBORB24004-100EA 0 CZK
BIRBORB24004-100
Anti-Influenza A H5 Mouse Monoclonal Antibody
Protilátky
Type: Primary
Antigen: Influenza A H5
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR