Tisk…

Anti-Toxoplasma gondii Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase))

Dodavatel: Biorbyt
BIRBORB24024-1EA 0 CZK
BIRBORB24024-1
Anti-Toxoplasma gondii Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase))
Protilátky
Type: Primary
Antigen: Toxoplasma gondii
Clonality: monoclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR