Tisk…

Anti-HBsAg (ayw) Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase))

Dodavatel: Biorbyt
BIRBORB24067-1EA 0 CZK
BIRBORB24067-1
Anti-HBsAg (ayw) Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase))
Protilátky
Type: Primary
Antigen: HBsAg (ayw)
Clonality: monoclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR