Tisk…

Anti-HCV NS-5a Mouse monoclonal antibody

Dodavatel: Biorbyt
BIRBORB24160-100EA 0 CZK
BIRBORB24160-100
Anti-HCV NS-5a Mouse monoclonal antibody
Protilátky
Western Blot: 1:1000; Immunochemistry: 1:20

Type: Primary
Antigen: HCV NS-5a
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR