Tisk…

Anti-Cholecystokinin Receptor A Rabbit Polyclonal Antibody

Dodavatel: Biorbyt
BIRBORB24176-100EA 0 CZK
BIRBORB24176-100
Anti-Cholecystokinin Receptor A Rabbit Polyclonal Antibody
Protilátky
Western Blot: 1:2000

Type: Primary
Antigen: Cholecystokinin Receptor A
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR