Tisk…

Anti-Calcitonin Rabbit Polyclonal Antibody

Dodavatel: Biorbyt
BIRBORB24179-20EA 0 CZK
BIRBORB24179-20
Anti-Calcitonin Rabbit Polyclonal Antibody
Protilátky
RIA: 1:15000

Type: Primary
Antigen: Calcitonin
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR