Tisk…

Anti-GHR Mouse monoclonal antibody

Dodavatel: Biorbyt
BIRBORB24269-1EA 0 CZK
BIRBORB24269-1
Anti-GHR Mouse monoclonal antibody
Protilátky
Type: Primary
Antigen: GHR (growth hormone receptor)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR